Learn More
Cayman Chemical OSU03012, Purity: ≥98%, CAS: 742112-33-0, 50 mg
SDP
Catalog No. 502888309
Encompass_Preferred
An analog of celecoxib that inhibits PDK-1 (IC50 = 5 µM), and therefore Akt activation, with no measurable COX-2 inhibition up to 50 µM; inhibits tumor cell growth (IC50s ~ 1.1 µM) across a panel of 60 cancer cell lines and induces apoptosis of chronic lymphocytic leukemia cells independent of bcl-2 overexpression using both caspase-dependent and independent pathways
Catalog No. 50-288-8309 Supplier Cayman Chemical Supplier No. 1000800550 MG
An analog of celecoxib that inhibits PDK-1 (IC50 = 5 µM), and therefore Akt activation, with no measurable COX-2 inhibition up to 50 µM; inhibits tumor cell growth (IC50s ~ 1.1 µM) across a panel of 60 cancer cell lines and induces apoptosis of chronic lymphocytic leukemia cells independent of bcl-2 overexpression using both caspase-dependent and independent pathways