Learn More
TARGETMOL CHEMICALS INC OT-551 HCL 5MG
SDP
Catalog No. 502580527
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. OT-551 HCl is an NF-(kappa)b inhibitor a disubstituted hydroxylamine with antioxidant properties can be used for the treatment of cataracts and age-related macular degeneration and geographic atrophy (GA). purity: 99%
Catalog No. 50-258-0527 Supplier TARGETMOL CHEMICALS INC Supplier No. T28273L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. OT-551 HCl is an NF-(kappa)b inhibitor a disubstituted hydroxylamine with antioxidant properties can be used for the treatment of cataracts and age-related macular degeneration and geographic atrophy (GA). purity: 99%