Learn More
Medchemexpress LLC HY-136241 10mg , OT-82 CAS:1800487-55-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13624110MG
Encompass_Preferred
HY-136241 10mg OT-82 CAS: 1800487-55-1 OT-82 is a potent, selective and orally active inhibitor of NAMPT. OT-82 is selectively toxic to cells of hematopoietic origin and induces cell death in a NAD+ dependent manner. OT-82 is a promising antineoplastic agent for the study of hematological malignancies. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0783