Learn More
Medchemexpress LLC OT-82 | 1800487-55-1 | 99.95% | 424.47 | 10 MG
SDP
Catalog No. 5000351767
Encompass_Preferred
OT-82 is a potent, selective, and orally active inhibitor of NAMPT. It is selectively toxic to cells of hematopoietic origin and induces cell death in a NAD+ dependent manner. It is a promising antineoplastic agent for the study of hematological malignancies.
  • Potent, selective, and orally active NAMPT inhibitor
  • Selectively toxic to cells of hematopoietic origin
  • Induces cell death in a NAD+ dependent manner
  • Promising antineoplastic agent
  • Demonstrates tissue-selective cytotoxicity
  • Inhibits recombinant NAMPT activity
  • Reduces cellular NAD and ATP concentrations
  • Activates caspase-3 and increases sub-G1 DNA content
  • Depolarizes the mitochondrial membrane
  • Increases survival in a SC xenograft model of Burkitt's lymphoma
  • Potently inhibits tumor growth in a multiple myeloma mouse model
Catalog No. 50-003-51767 Supplier Medchemexpress LLC Supplier No. HY13624110MM/1ML
OT-82 is a potent, selective, and orally active inhibitor of NAMPT. It is selectively toxic to cells of hematopoietic origin and induces cell death in a NAD+ dependent manner. It is a promising antineoplastic agent for the study of hematological malignancies.
  • Potent, selective, and orally active NAMPT inhibitor
  • Selectively toxic to cells of hematopoietic origin
  • Induces cell death in a NAD+ dependent manner
  • Promising antineoplastic agent
  • Demonstrates tissue-selective cytotoxicity
  • Inhibits recombinant NAMPT activity
  • Reduces cellular NAD and ATP concentrations
  • Activates caspase-3 and increases sub-G1 DNA content
  • Depolarizes the mitochondrial membrane
  • Increases survival in a SC xenograft model of Burkitt's lymphoma
  • Potently inhibits tumor growth in a multiple myeloma mouse model