Learn More
Medchemexpress LLC OT-82 | 1800487-55-1 | 99.95% | 424.47 | 25 MG
SDP
Catalog No. 5000347850
Encompass_Preferred
OT-82 is a potent, selective, and orally active inhibitor of NAMPT. It is selectively toxic to cells of hematopoietic origin and induces cell death in a NAD+-dependent manner. OT-82 is a promising antineoplastic agent for the study of hematological malignancies.
  • Potent, selective, and orally active NAMPT inhibitor.
  • Selectively toxic to cells of hematopoietic origin.
  • Induces cell death in a NAD+-dependent manner.
  • Demonstrates tissue-selective and cancer-selective cytotoxicity.
  • Inhibits recombinant NAMPT activity and reduces cellular NAD and ATP concentrations.
  • Activates caspase-3, increases sub-G1 DNA content, and depolarizes mitochondrial membrane.
  • Stable for 3 years at -20°C or 2 years at 4°C as powder.
  • Stable for 2 years at -80°C or 1 year at -20°C when in solvent.
Catalog No. 50-003-47850 Supplier Medchemexpress LLC Supplier No. 25MGHY136241
OT-82 is a potent, selective, and orally active inhibitor of NAMPT. It is selectively toxic to cells of hematopoietic origin and induces cell death in a NAD+-dependent manner. OT-82 is a promising antineoplastic agent for the study of hematological malignancies.
  • Potent, selective, and orally active NAMPT inhibitor.
  • Selectively toxic to cells of hematopoietic origin.
  • Induces cell death in a NAD+-dependent manner.
  • Demonstrates tissue-selective and cancer-selective cytotoxicity.
  • Inhibits recombinant NAMPT activity and reduces cellular NAD and ATP concentrations.
  • Activates caspase-3, increases sub-G1 DNA content, and depolarizes mitochondrial membrane.
  • Stable for 3 years at -20°C or 2 years at 4°C as powder.
  • Stable for 2 years at -80°C or 1 year at -20°C when in solvent.