Learn More
Selleck Chemical LLC OT-82-5mg
SDP
Catalog No. 502389655
Encompass_Preferred
OT-82 is a novel nicotinamide phosphoribosyltransferase (NAMPT) inhibitor with average IC50s of 13.03   nM in non-HP cancer cells and 2.89   nM in HP cancer cells, respectively.
Catalog No. 50-238-9655 Supplier Selleck Chemical LLC Supplier No. S97065MG
OT-82 is a novel nicotinamide phosphoribosyltransferase (NAMPT) inhibitor with average IC50s of 13.03   nM in non-HP cancer cells and 2.89   nM in HP cancer cells, respectively.