Learn More
BURKLE INC OTAL battery foot pump, PP/PVC, tube Ø 0.47 in. (12 mm)
Catalog No. 5000025961
Encompass_Preferred
The OTAL battery pump is a flexible pump with hose, tap and dispensing nozzle. Unlike conventional pumps, there is no need to place or hold the container under the discharge elbow. Instead, the flexible 1.5 meter (4.92 ft.) hose can simply be placed on the container, allowing greater flexibility in positioning the container.
The OTAL battery pump is equipped with a flexible PVC discharge hose, a PP/PE stopcock, and a PE dispensing nozzle. The delivery tubes are made of PP. The discharge hose can be shortened or replaced if necessary.
The stopcock allows good dosing. It also prevents the liquid from running on.
The pump is particularly suitable for filling car batteries with acid or distilled water.
Catalog No. 50-000-25961 Supplier BURKLE INC Supplier No. 50006000
The OTAL battery pump is a flexible pump with hose, tap and dispensing nozzle. Unlike conventional pumps, there is no need to place or hold the container under the discharge elbow. Instead, the flexible 1.5 meter (4.92 ft.) hose can simply be placed on the container, allowing greater flexibility in positioning the container.
The OTAL battery pump is equipped with a flexible PVC discharge hose, a PP/PE stopcock, and a PE dispensing nozzle. The delivery tubes are made of PP. The discharge hose can be shortened or replaced if necessary.
The stopcock allows good dosing. It also prevents the liquid from running on.
The pump is particularly suitable for filling car batteries with acid or distilled water.