Learn More
BURKLE INC OTAL disposable foot pump, delivery tubes PE
Catalog No. 5000025611
Encompass_Preferred
The OTAL foot pump with disposable HDPE delivery tubes offers a hygienic, contamination-free solution for filling containers up to 220 liters (58 gallons). The large-volume, foot-operated bellows pump makes transferring liquids effortless, with the fluid only contacting the easily replaceable disposable delivery tube.
Its compact, space-saving design and single-use delivery tube eliminate the need for time-consuming cleaning and prevent cross-contamination — perfect for sensitive or varying fluids. The adjustable tube can be cut to fit container heights and ensures complete emptying. Compatible with airtight openings ranging from 1.2 to 2.75 inches (30 to 70 mm).
The universal PVC plug fits containers without threading, ideal for drums, cans, and barrels up to 35 inches (90 cm) tall. The OTAL disposable foot pump guarantees clean, safe, and efficient filling every time.
Catalog No. 50-000-25611 Supplier BURKLE INC Supplier No. 50008012
The OTAL foot pump with disposable HDPE delivery tubes offers a hygienic, contamination-free solution for filling containers up to 220 liters (58 gallons). The large-volume, foot-operated bellows pump makes transferring liquids effortless, with the fluid only contacting the easily replaceable disposable delivery tube.
Its compact, space-saving design and single-use delivery tube eliminate the need for time-consuming cleaning and prevent cross-contamination — perfect for sensitive or varying fluids. The adjustable tube can be cut to fit container heights and ensures complete emptying. Compatible with airtight openings ranging from 1.2 to 2.75 inches (30 to 70 mm).
The universal PVC plug fits containers without threading, ideal for drums, cans, and barrels up to 35 inches (90 cm) tall. The OTAL disposable foot pump guarantees clean, safe, and efficient filling every time.