Learn More
BURKLE INC OTAL foot pump PP, tube Ø 0.59 in. (15 mm), pump cap. 5.28 gal. (20l)/min
Catalog No. 5000025755
Encompass_Preferred
The OTAL foot pump with PP delivery tubes is a reliable, flexible and cost-effective pump for filling liquids.
Various delivery tubes with different diameters and flow rates are available to meet specific requirements.
The PP delivery tubes offer good resistance to weak acids, alkalis and detergents. The outlet can be turned up to prevent dripping.
The OTAL foot pump with PP delivery tubes is easy to use and durable. It is the ideal choice for cost-effective and reliable fluid transfer.
Catalog No. 50-000-25755 Supplier BURKLE INC Supplier No. 50001500
The OTAL foot pump with PP delivery tubes is a reliable, flexible and cost-effective pump for filling liquids.
Various delivery tubes with different diameters and flow rates are available to meet specific requirements.
The PP delivery tubes offer good resistance to weak acids, alkalis and detergents. The outlet can be turned up to prevent dripping.
The OTAL foot pump with PP delivery tubes is easy to use and durable. It is the ideal choice for cost-effective and reliable fluid transfer.