Learn More
BURKLE INC OTAL foot pump PVDF, tube Ø 0.63 in. (16 mm), pump cap. 5.28 gal. (20l)/min
Catalog No. 5000026105
Encompass_Preferred
The OTAL foot pump with PVDF delivery tubes is a reliable pump with high chemical resistance.
The special properties of PVDF make the foot pump resistant to chlorine bleach and other chemicals. PVDF offers even better resistance to various acids and alkalis than other materials such as PP.
The pump outlet can be turned up to prevent dripping.
The OTAL foot pump PVDF is easy to use and can transfer liquids quickly and efficiently. It is an economical and practical solution for many applications.
Catalog No. 50-000-26105 Supplier BURKLE INC Supplier No. 50004016
The OTAL foot pump with PVDF delivery tubes is a reliable pump with high chemical resistance.
The special properties of PVDF make the foot pump resistant to chlorine bleach and other chemicals. PVDF offers even better resistance to various acids and alkalis than other materials such as PP.
The pump outlet can be turned up to prevent dripping.
The OTAL foot pump PVDF is easy to use and can transfer liquids quickly and efficiently. It is an economical and practical solution for many applications.