Learn More
BURKLE INC OTAL foot pump V2A, tube Ø 0.71 in. (18 mm), pump cap. 6.87 gal. (26l)/min
Catalog No. 5000026107
Encompass_Preferred
The OTAL foot pump with delivery tubes made of high-quality stainless steel 1.4301 (304) is ideal for bottling liquids of hazard class AIII (VbF) that are difficult to ignite. The stainless steel delivery tubes guarantee high chemical resistance as well as high temperature resistance.
Compared to models made of plastic, stainless steel delivery tubes are particularly robust, dimensionally stable, resilient and extremely durable.
Catalog No. 50-000-26107 Supplier BURKLE INC Supplier No. 50005018
The OTAL foot pump with delivery tubes made of high-quality stainless steel 1.4301 (304) is ideal for bottling liquids of hazard class AIII (VbF) that are difficult to ignite. The stainless steel delivery tubes guarantee high chemical resistance as well as high temperature resistance.
Compared to models made of plastic, stainless steel delivery tubes are particularly robust, dimensionally stable, resilient and extremely durable.