Learn More
BURKLE INC OTAL hand pump V2A, tube Ø 0.71 in. (18 mm), pump cap. 6.87 gal. (26l)/min
Catalog No. 5000025812
Encompass_Preferred
The OTAL hand pump with delivery tubes made of high-quality stainless steel 1.4301 (304) is ideal for filling difficult to ignite liquids of hazard class AIII (VbF). The stainless steel delivery tubes guarantee high chemical resistance as well as high temperature resistance.
Compared to plastic models, stainless steel delivery tubes are particularly robust, dimensionally stable, resilient and extremely durable.
Catalog No. 50-000-25812 Supplier BURKLE INC Supplier No. 50054000
The OTAL hand pump with delivery tubes made of high-quality stainless steel 1.4301 (304) is ideal for filling difficult to ignite liquids of hazard class AIII (VbF). The stainless steel delivery tubes guarantee high chemical resistance as well as high temperature resistance.
Compared to plastic models, stainless steel delivery tubes are particularly robust, dimensionally stable, resilient and extremely durable.