Learn More
BURKLE INC OTAL hand pump PP, tube Ø 0.71 in. (18 mm), pump cap. 6.87 gal. (26l)/min
Catalog No. 5000025759
Encompass_Preferred
The OTAL hand pump with delivery tubes made of PP is a reliable, flexible and inexpensive pump for filling liquids.
To meet specific requirements, various delivery tubes with different diameters and flow rates are available. The delivery tubes made of PP offer good resistance to weak acids, alkalis and cleaning agents. The outlet can be turned upwards to prevent dripping.
The OTAL hand pump with PP delivery tubes is easy to use and durable. It is the ideal choice for cost-effective and reliable filling of liquids.
Catalog No. 50-000-25759 Supplier BURKLE INC Supplier No. 50051800
The OTAL hand pump with delivery tubes made of PP is a reliable, flexible and inexpensive pump for filling liquids.
To meet specific requirements, various delivery tubes with different diameters and flow rates are available. The delivery tubes made of PP offer good resistance to weak acids, alkalis and cleaning agents. The outlet can be turned upwards to prevent dripping.
The OTAL hand pump with PP delivery tubes is easy to use and durable. It is the ideal choice for cost-effective and reliable filling of liquids.