Learn More
BURKLE INC OTAL hand pump hose & stopcock, PP/PVC,tube Ø 0.47 in. (12 mm)
Catalog No. 5000026026
Encompass_Preferred
The OTAL hand pump with hose and stopcock is a flexible and space-saving pump. Unlike conventional pumps, the container does not have to be placed or held under the discharge elbow. Instead, the flexible 1.5 meter (4.92 ft.) hose can simply be routed to the container, allowing greater flexibility in positioning the container.
The OTAL hand pump is equipped with a flexible discharge hose made of PVC and a stopcock made of PP/PE. The delivery tubes are made of PP. The discharge hose can be shortened or replaced if necessary.
The stopcock enables good dosing. It also prevents the liquid from running on unintentionally.
Catalog No. 50-000-26026 Supplier BURKLE INC Supplier No. 50056000
The OTAL hand pump with hose and stopcock is a flexible and space-saving pump. Unlike conventional pumps, the container does not have to be placed or held under the discharge elbow. Instead, the flexible 1.5 meter (4.92 ft.) hose can simply be routed to the container, allowing greater flexibility in positioning the container.
The OTAL hand pump is equipped with a flexible discharge hose made of PVC and a stopcock made of PP/PE. The delivery tubes are made of PP. The discharge hose can be shortened or replaced if necessary.
The stopcock enables good dosing. It also prevents the liquid from running on unintentionally.