Learn More
BURKLE INC OTAL hand pump PVDF, tube Ø 0.63 in. (16 mm), pump cap. 5.28 gal. (20l)/min
Catalog No. NC2187658
Encompass_Preferred
The OTAL hand pump with PVDF delivery tubes is a reliable pump with high chemical resistance.
Due to the special properties of PVDF, the hand pump is resistant to chlorine bleach and other chemicals. PVDF offers even better resistance to various acids and alkalis than other materials such as PP.
The pump outlet can be turned up to prevent dripping.
The OTAL PVDF hand pump is easy to use and can transfer liquids quickly and efficiently. It is an economical and practical solution for many applications.
This item is not returnable. View return policy
Catalog No. NC2187658 Supplier BURKLE INC Supplier No. 50053000
The OTAL hand pump with PVDF delivery tubes is a reliable pump with high chemical resistance.
Due to the special properties of PVDF, the hand pump is resistant to chlorine bleach and other chemicals. PVDF offers even better resistance to various acids and alkalis than other materials such as PP.
The pump outlet can be turned up to prevent dripping.
The OTAL PVDF hand pump is easy to use and can transfer liquids quickly and efficiently. It is an economical and practical solution for many applications.