Learn More
BURKLE INC OTAL mini hand pump, PP, tube Ø 0.39 in. (10 mm), 1.06 gal. (4l)/min
Catalog No. 5000025676
Encompass_Preferred
The OTAL mini hand pump PP is ideal for use in smaller containers. With its smaller plug, it is designed for container openings from 20 to 36 mm (0.79 to 1.42 in.). The pump has a maximum immersion depth of 45 cm (17.72 in.) and is therefore ideal for use in small containers up to approx. 10 liters (2.64 gal.).
The delivery tubes of the hand pump are made of PP, the universal plug is made of soft PVC. The OTAL Mini hand pump is resistant to weak acids and alkalis.
The pump outlet can be turned up to prevent dripping.
The OTAL mini is an economical alternative to comparable models and offers reliable filling from small containers.
Catalog No. 50-000-25676 Supplier BURKLE INC Supplier No. 50051000
The OTAL mini hand pump PP is ideal for use in smaller containers. With its smaller plug, it is designed for container openings from 20 to 36 mm (0.79 to 1.42 in.). The pump has a maximum immersion depth of 45 cm (17.72 in.) and is therefore ideal for use in small containers up to approx. 10 liters (2.64 gal.).
The delivery tubes of the hand pump are made of PP, the universal plug is made of soft PVC. The OTAL Mini hand pump is resistant to weak acids and alkalis.
The pump outlet can be turned up to prevent dripping.
The OTAL mini is an economical alternative to comparable models and offers reliable filling from small containers.