Learn More
Medchemexpress LLC Otaplimastat (SP-8203) | 1176758-04-5 | 98.11% | 534.61 | 50 MG
SDP
Catalog No. 5000344263
Encompass_Preferred
Otaplimastat (SP-8203) is a matrix metalloproteinase (MMP) inhibitor that competitively blocks N-methyl-D-aspartate (NMDA) receptor-mediated excitotoxicity. It also exhibits anti-oxidant activity and can be used for research on brain ischemic injury.
  • MMP inhibitor
  • Blocks NMDA receptor-mediated excitotoxicity competitively
  • Exhibits anti-oxidant activity
  • Potential for research of brain ischemic injury
  • Protects neuronal cells against NMDA-induced cell death
  • Inhibits Ca2+ influx following activation of NMDA receptors in primary cultured neurons
  • Suppresses H2O2-induced cell death and reactive oxygen species production
  • Prevents ischemic neuronal death in the occlusion model of MCA in rats
  • Attenuates impairment of stroke-induced motor function in rats
Catalog No. 50-003-44263 Supplier Medchemexpress LLC Supplier No. HY10909750MG
Otaplimastat (SP-8203) is a matrix metalloproteinase (MMP) inhibitor that competitively blocks N-methyl-D-aspartate (NMDA) receptor-mediated excitotoxicity. It also exhibits anti-oxidant activity and can be used for research on brain ischemic injury.
  • MMP inhibitor
  • Blocks NMDA receptor-mediated excitotoxicity competitively
  • Exhibits anti-oxidant activity
  • Potential for research of brain ischemic injury
  • Protects neuronal cells against NMDA-induced cell death
  • Inhibits Ca2+ influx following activation of NMDA receptors in primary cultured neurons
  • Suppresses H2O2-induced cell death and reactive oxygen species production
  • Prevents ischemic neuronal death in the occlusion model of MCA in rats
  • Attenuates impairment of stroke-induced motor function in rats