Learn More
Aviva Systems Biology Corp OTC Antibody - N-terminal region (ARP41766_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41766_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against OTC. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human OTC. Alternate names: OCTDApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-53314