Learn More
Aviva Systems Biology Corp Otc Antibody - C-terminal region (ARP41767_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41767_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Otc. It was validated on Western Blot. . Alternate names: AI265390, Sf, spfApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-284-954