Learn More
ABclonal Technology OTC Rabbit mAb
SDP
Catalog No. 5000008671
Encompass_Preferred
This nuclear gene encodes a mitochondrial matrix enzyme The encoded protein is involved in the urea cycle which functions to detoxify ammonia into urea for excretion Mutations in this enzyme lead to ornithine transcarbamylase deficiency which causes hyperammonemia
Catalog No. 50-000-08671 Supplier ABclonal Technology Supplier No. A2501520UL
This nuclear gene encodes a mitochondrial matrix enzyme The encoded protein is involved in the urea cycle which functions to detoxify ammonia into urea for excretion Mutations in this enzyme lead to ornithine transcarbamylase deficiency which causes hyperammonemia