Learn More
Biolog Inc OTEB - Oral Treponeme Enrichment Broth, 10 Tubes/Pkg
SDP
Catalog No. NC1244254
Encompass
This item has a minimum qty of 2 per supplier requirements.
Oral Treponeme Enrichment Broth (OTEB) is an enriched nonselective medium used for the isolation, cultivation, and maintenance of Treponema spp. from clinical specimens. This medium is prepared, dispensed, and packaged under oxygen-free conditions to prevent the formation of oxidized products prior to use.OTEB is a complex peptone yeast extract medium containing volatile fatty acids and calf serum. Volatile fatty acids and serum are necessary growth factors for the recovery of Treponemes from clinical specimens. A variety of sugars have been added as a source of carbohydrates. The addition of short-chain fatty acids (SCFA) and serum may also aid in the recovery and growth of fastideous organisms with complex nutritional requirements.
Catalog No. NC1244254 Supplier Biolog Inc Supplier No. AS603
This item has a minimum qty of 2 per supplier requirements.
Oral Treponeme Enrichment Broth (OTEB) is an enriched nonselective medium used for the isolation, cultivation, and maintenance of Treponema spp. from clinical specimens. This medium is prepared, dispensed, and packaged under oxygen-free conditions to prevent the formation of oxidized products prior to use.OTEB is a complex peptone yeast extract medium containing volatile fatty acids and calf serum. Volatile fatty acids and serum are necessary growth factors for the recovery of Treponemes from clinical specimens. A variety of sugars have been added as a source of carbohydrates. The addition of short-chain fatty acids (SCFA) and serum may also aid in the recovery and growth of fastideous organisms with complex nutritional requirements.