Learn More
Selleck Chemical LLC Otenabant
SDP
Catalog No. 5000001470
Encompass_Preferred
Otenabant (CP-945598) HCl is a potent and selective cannabinoid receptor CB1 antagonist with Ki of 0 7 nM exhibits 10 000-fold greater selectivity against human CB2 receptor Phase 1
Catalog No. 50-000-01470 Supplier Selleck Chemical LLC Supplier No. S801210MG
Otenabant (CP-945598) HCl is a potent and selective cannabinoid receptor CB1 antagonist with Ki of 0 7 nM exhibits 10 000-fold greater selectivity against human CB2 receptor Phase 1