Learn More
Medchemexpress LLC HY-10871 5mg Medchemexpress, Otenabant CAS:686344-29-6 Purity:>98%
SDP
Catalog No. 501968619
Encompass_Preferred
Medchemexpress, HY-10871 5mg Otenabant CAS:686344-29-6 Otenabant is a potent and selective cannabinoid receptor CB1 antagonist with Ki of 0.7 nM, exhibits 10,000-fold greater selectivity against human CB2 receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8619 Supplier Medchemexpress LLC Supplier No. HY108715MG
Medchemexpress, HY-10871 5mg Otenabant CAS:686344-29-6 Otenabant is a potent and selective cannabinoid receptor CB1 antagonist with Ki of 0.7 nM, exhibits 10,000-fold greater selectivity against human CB2 receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.