Learn More
Medchemexpress LLC HY-101381 1mg Medchemexpress, Otenzepad CAS:102394-31-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1013811MG
Encompass_Preferred
Medchemexpress, HY-101381 1mg Otenzepad CAS:102394-31-0 Otenzepad (AF-DX 116) is a selective and competitive M2 muscarinic acetylcholine receptor antagonist, with IC50 values of 640 nM and 386 nM for rabbit peripheral lung and rat heart, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7839