Learn More
TARGETMOL CHEMICALS INC OTESECONAZOLE 5MG
SDP
Catalog No. 502581178
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Oteseconazole (VT-1161) is an orally active anti-fungal agent. Oteseconazole displays no obvious effect on human CYP51. It also potently binds to and inhibits Candida albicans CYP51 (Kd <39 nM). purity: 99%
Catalog No. 50-258-1178 Supplier TARGETMOL CHEMICALS INC Supplier No. T164125MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Oteseconazole (VT-1161) is an orally active anti-fungal agent. Oteseconazole displays no obvious effect on human CYP51. It also potently binds to and inhibits Candida albicans CYP51 (Kd <39 nM). purity: 99%