Learn More
Medchemexpress LLC HY-17643 5mg Medchemexpress, Oteseconazole CAS:1340593-59-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY176435MG
Encompass_Preferred
Medchemexpress, HY-17643 5mg Oteseconazole CAS:1340593-59-0 Oteseconazole (VT-1161) is an orally active anti-fungal agent, potently binds to and inhibits Candida albicans CYP51 (Kd, <39 nM), shows no obvious effect on human CYP51. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0001