Learn More
Med Vet International Motazol Otic Suspension, 215gm, Each
Catalog No. 502828822
Encompass_Preferred
MotaZol Otic Suspension is indicated for the treatment of otitis externa in dogs caused by susceptible strains of yeast (Malassezia pachydermatis) and bacteria (Pseudomonas spp. [including P. aeruginosa], coagulase-positive staphylococci, Enterococcus faecalis, Proteus mirabilis, and beta-hemolytic streptococci).
Catalog No. 50-282-8822 Supplier Med Vet International Supplier No. RXMOTA215215V
MotaZol Otic Suspension is indicated for the treatment of otitis externa in dogs caused by susceptible strains of yeast (Malassezia pachydermatis) and bacteria (Pseudomonas spp. [including P. aeruginosa], coagulase-positive staphylococci, Enterococcus faecalis, Proteus mirabilis, and beta-hemolytic streptococci).