Learn More
TARGETMOL CHEMICALS INC OTILONIUM BROMIDE 100MG
SDP
Catalog No. 502577378
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Otilonium bromide (SP63) an antimuscarinic is utilized as a spasmolytic agent. purity: 99%
Catalog No. 50-257-7378 Supplier TARGETMOL CHEMICALS INC Supplier No. T1474100MG
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Otilonium bromide (SP63) an antimuscarinic is utilized as a spasmolytic agent. purity: 99%