Learn More
Medchemexpress LLC Otilonium (bromide) | 26095-59-0 | 99.7% | 1 ML
SDP
Catalog No. 5000408030
Encompass_Preferred
Otilonium bromide is an orally active mAChR inhibitor and smooth muscle relaxant. It can interfere with the mobilization of calcium in intestinal smooth muscle and is used for research on irritable bowel syndrome.
  • Inhibits acetylcholine-induced human colonic crypt calcium signals in a dose-dependent manner with an IC50 of 880 nM.
  • Inhibits Ca2+ uptake in bovine chromaffin cells at 30 μM.
  • Targets mAChR and is involved in GPCR/G Protein and Neuronal Signaling pathways.
Catalog No. 50-004-08030 Supplier Medchemexpress LLC Supplier No. HYB0499A10MM/1ML
Otilonium bromide is an orally active mAChR inhibitor and smooth muscle relaxant. It can interfere with the mobilization of calcium in intestinal smooth muscle and is used for research on irritable bowel syndrome.
  • Inhibits acetylcholine-induced human colonic crypt calcium signals in a dose-dependent manner with an IC50 of 880 nM.
  • Inhibits Ca2+ uptake in bovine chromaffin cells at 30 μM.
  • Targets mAChR and is involved in GPCR/G Protein and Neuronal Signaling pathways.