Learn More
Medchemexpress LLC Otilonium bromide | 26095-59-0 | MFCD00079361 | 99.7% | 50 MG
SDP
Catalog No. 5000409457
Encompass_Preferred
Otilonium bromide (OB) is an orally active mAChR inhibitor and smooth muscle relaxant. It can interfere with the mobilization of calcium in intestinal smooth muscle and is used for research of irritable bowel syndrome.
  • Orally active mAChR inhibitor
  • Smooth muscle relaxant
  • Interferes with calcium mobilization in intestinal smooth muscle
  • Used for research of irritable bowel syndrome
Catalog No. 50-004-09457 Supplier Medchemexpress LLC Supplier No. HYB0499A50MG
Otilonium bromide (OB) is an orally active mAChR inhibitor and smooth muscle relaxant. It can interfere with the mobilization of calcium in intestinal smooth muscle and is used for research of irritable bowel syndrome.
  • Orally active mAChR inhibitor
  • Smooth muscle relaxant
  • Interferes with calcium mobilization in intestinal smooth muscle
  • Used for research of irritable bowel syndrome