Learn More
Aviva Systems Biology Corp OTOP1 Antibody - C-terminal region (ARP65798_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65798_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OTOP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human OTOP1. Alternate names: MGC163302, MGC163304ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-574