Learn More
Aviva Systems Biology Corp OTP Antibody - N-terminal region (ARP34633_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34633_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OTP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human OTP. Alternate names: MGC3161ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55492