Learn More
Aviva Systems Biology Corp OTP Antibody - C-terminal region (ARP39679_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39679_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OTP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human OTP. Alternate names: ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-53149