Learn More
Medchemexpress LLC OTs-C6-OBn | 126519-80-0 | 98.1% | 500 MG
SDP
Catalog No. 5000340356
Encompass_Preferred
OTs-C6-OBn is an alkyl chain-based PROTAC linker utilized in the synthesis of PROTAC SGK3 degrader-1 (HY-125878). PROTACs, or Proteolysis Targeting Chimeras, consist of two distinct ligands connected by a linker: one ligand binds to an E3 ubiquitin ligase, and the other binds to a target protein. These molecules function by exploiting the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTAC SGK3 degrader-1
  • PROTACs utilize the ubiquitin-proteasome system for selective protein degradation
Catalog No. 50-003-40356 Supplier Medchemexpress LLC Supplier No. HY130621500MG
OTs-C6-OBn is an alkyl chain-based PROTAC linker utilized in the synthesis of PROTAC SGK3 degrader-1 (HY-125878). PROTACs, or Proteolysis Targeting Chimeras, consist of two distinct ligands connected by a linker: one ligand binds to an E3 ubiquitin ligase, and the other binds to a target protein. These molecules function by exploiting the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTAC SGK3 degrader-1
  • PROTACs utilize the ubiquitin-proteasome system for selective protein degradation