Learn More
Pyramex™ OTS™ Safety Eyewear
Catalog No. 19155560 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Amber
IR/UV 3.0
IR/UV 5.0
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-155-560 IR/UV 5.0
19-155-561 IR/UV 3.0
19-155-559 Amber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-155-560 Supplier Pyramex™ Supplier No. S3550SFJ
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed to better fit today's prescription eye wear
  • Lightweight nylon temples adjust for length and pitch
  • Single lens encapsulates the eye for superb protection
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Fits over frames up to 5 3/8 in. W x 1 3/4 in. H (137 mm x 44 mm)
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Certifications/Compliance (Europe) EN-166
    Lens Tint IR/UV 5.0
    Frame Color Black
    Weight (Metric) 33 g
    Lens Material Polycarbonate