Learn More
Medchemexpress LLC OTS186935 hydrochloride | 99.6% | 522.31 | 50 MG
SDP
Catalog No. 5000437302
Encompass_Preferred
OTS186935 hydrochloride is a potent protein methyltransferase SUV39H2 inhibitor with an IC50 of 6.49 nM. It significantly inhibits tumor growth in mouse xenograft models without detectable toxicity and regulates the production of γ-H2AX in cancer cells.
  • Potent protein methyltransferase SUV39H2 inhibitor with an IC50 of 6.49 nM.
  • Shows significant inhibition of tumor growth in mouse xenograft models.
  • Does not exhibit any detectable toxicity in mouse xenograft models.
  • Regulates the production of γ-H2AX in cancer cells.
  • Inhibits A549 cell growth with an IC50 of 0.67 μM.
Catalog No. 50-004-37302 Supplier Medchemexpress LLC Supplier No. HY122181B50MG
OTS186935 hydrochloride is a potent protein methyltransferase SUV39H2 inhibitor with an IC50 of 6.49 nM. It significantly inhibits tumor growth in mouse xenograft models without detectable toxicity and regulates the production of γ-H2AX in cancer cells.
  • Potent protein methyltransferase SUV39H2 inhibitor with an IC50 of 6.49 nM.
  • Shows significant inhibition of tumor growth in mouse xenograft models.
  • Does not exhibit any detectable toxicity in mouse xenograft models.
  • Regulates the production of γ-H2AX in cancer cells.
  • Inhibits A549 cell growth with an IC50 of 0.67 μM.