Learn More
Selleck Chemical LLC OTS514 hydrochloride
SDP
Catalog No. 5000000946
Encompass_Preferred
OTS514 is a highly potent TOPK(T-LAK cell-originated protein kinase) inhibitor with an IC50 value of 2 6 nM OTS514 induces cell cycle arrest and apoptosis
Catalog No. 50-000-00946 Supplier Selleck Chemical LLC Supplier No. S76525MG
Add to Cart
Add to Cart
OTS514 is a highly potent TOPK(T-LAK cell-originated protein kinase) inhibitor with an IC50 value of 2 6 nM OTS514 induces cell cycle arrest and apoptosis