Learn More
Selleck Chemical LLC OTS514 hydrochloride-2mg
SDP
Catalog No. 502389770
Encompass_Preferred
OTS514 is a highly potent TOPK(T-LAK cell-originated protein kinase) inhibitor with an IC50 value of 2.6 nM. OTS514 induces cell cycle arrest and apoptosis.
Catalog No. 50-238-9770 Supplier Selleck Chemical LLC Supplier No. S76522MG
Add to Cart
Add to Cart
OTS514 is a highly potent TOPK(T-LAK cell-originated protein kinase) inhibitor with an IC50 value of 2.6 nM. OTS514 induces cell cycle arrest and apoptosis.