Learn More
TARGETMOL CHEMICALS INC OTS514 HYDROCHLORIDE 5MG
SDP
Catalog No. 502579935
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. OTS514 hydrochloride is a highly effective TOPK(T-LAK cell-originated protein kinase) inhibitor (IC50 2.6 nM). purity: 99%
Catalog No. 50-257-9935 Supplier TARGETMOL CHEMICALS INC Supplier No. T41345MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. OTS514 hydrochloride is a highly effective TOPK(T-LAK cell-originated protein kinase) inhibitor (IC50 2.6 nM). purity: 99%