Learn More
Medchemexpress LLC HY-12467 10mg Medchemexpress, OTS964 (hydrochloride) CAS:1338545-07-5 Purity:>98%
SDP
Catalog No. 502028882
Encompass_Preferred
Medchemexpress, HY-12467 10mg OTS964 (hydrochloride) CAS:1338545-07-5 OTS964 hydrochloride is an orally active, high affinity and selective TOPK (T-lymphokine-activated killer cell-originated protein kinase) inhibitor with an IC50 of 28 nM. OTS964 hydrochloride is also a potent inhibitor of the cyclin-dependent kinase CDK11, which binds to CDK11B with a Kd of 40 nM[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8882 Supplier Medchemexpress LLC Supplier No. HY1246710MG
Medchemexpress, HY-12467 10mg OTS964 (hydrochloride) CAS:1338545-07-5 OTS964 hydrochloride is an orally active, high affinity and selective TOPK (T-lymphokine-activated killer cell-originated protein kinase) inhibitor with an IC50 of 28 nM. OTS964 hydrochloride is also a potent inhibitor of the cyclin-dependent kinase CDK11, which binds to CDK11B with a Kd of 40 nM[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.