Learn More
Cayman Chemical OTSSP167 (hydrochloride), Purity: ≥95%, CAS: 1431698-10-0, 10 mg
SDP
Catalog No. 502967037
Encompass_Preferred
A potent inhibitor of MELK (IC50 = 0.41 nM); blocks the phosphorylation of MELK-specific substrates and reduces the ability of MCF-7 breast cancer cells to invade and form spheroids in Matrigel; suppresses the growth of xenograft tumors in mice
Catalog No. 50-296-7037 Supplier Cayman Chemical Supplier No. 1687310 MG
A potent inhibitor of MELK (IC50 = 0.41 nM); blocks the phosphorylation of MELK-specific substrates and reduces the ability of MCF-7 breast cancer cells to invade and form spheroids in Matrigel; suppresses the growth of xenograft tumors in mice