Learn More
TARGETMOL CHEMICALS INC OTSSP167 10MG
SDP
Catalog No. 502577308
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. OTSSP167 (OTS167) is an orally available inhibitor of maternal embryonic leucine zipper kinase (MELK) with potential antineoplastic activity. purity: 99%
Catalog No. 50-257-7308 Supplier TARGETMOL CHEMICALS INC Supplier No. T175910MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. OTSSP167 (OTS167) is an orally available inhibitor of maternal embryonic leucine zipper kinase (MELK) with potential antineoplastic activity. purity: 99%