Learn More
Medchemexpress LLC HY-15512A 5mg , OTSSP167 (hydrochloride) CAS:1431698-10-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15512A5MG
Encompass_Preferred
HY-15512A 5mg OTSSP167 (hydrochloride) CAS: 1431698-10-0 OTSSP167 (OTS167) hydrochloride is a highly potent and ATP-competitive MELK inhibitor with IC50 value of 0.41 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0918