Learn More
TARGETMOL CHEMICALS INC OTSSP167 HYDROCHLORIDE 10MG
SDP
Catalog No. 502577213
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. OTSSP167 hydrochloride is a highly potent inhibitor of MELK(IC50 0.41 nM). purity: 99%
Catalog No. 50-257-7213 Supplier TARGETMOL CHEMICALS INC Supplier No. T2155910MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. OTSSP167 hydrochloride is a highly potent inhibitor of MELK(IC50 0.41 nM). purity: 99%