Learn More
Waters Corp Waters Otto Cartridge Startup Kit, 1 cc, for 13 mm Tubes
Catalog No. 502390885 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit provides essential components for starting solid phase extraction (SPE) applications with Otto systems. It is designed for use with 13 mm collection tubes and includes cartridges, racks, and waste collection.
  • Includes 24 flanged or flangeless 1 cc cartridges
  • Features a 1 cc top rack
  • Equipped with a 13 mm tube collection rack for 100 mm tubes
  • Comes with a tube spacer for 75 mm tubes
  • Includes waste collection components
Catalog No. 50-239-0885 Supplier Waters Corp Supplier No. 176004606
This kit provides essential components for starting solid phase extraction (SPE) applications with Otto systems. It is designed for use with 13 mm collection tubes and includes cartridges, racks, and waste collection.
  • Includes 24 flanged or flangeless 1 cc cartridges
  • Features a 1 cc top rack
  • Equipped with a 13 mm tube collection rack for 100 mm tubes
  • Comes with a tube spacer for 75 mm tubes
  • Includes waste collection components