Learn More
Waters Corp Waters Tube Collection Rack, 13 mm, for 6 cc Cartridges
Catalog No. 502392081 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tube collection rack serves as a crucial component of the Otto SPEcialist Positive Pressure Manifold system. It is engineered to facilitate sample collection by securely holding 13 mm tubes, specifically when utilized with 6 cc cartridges in laboratory applications.
  • Designed for use with the Otto SPEcialist Positive Pressure Manifold.
  • Accommodates 13 mm collection tubes.
  • Optimized for use with 6 cc cartridges.
Catalog No. 50-239-2081 Supplier Waters Corp Supplier No. 186009723
This tube collection rack serves as a crucial component of the Otto SPEcialist Positive Pressure Manifold system. It is engineered to facilitate sample collection by securely holding 13 mm tubes, specifically when utilized with 6 cc cartridges in laboratory applications.
  • Designed for use with the Otto SPEcialist Positive Pressure Manifold.
  • Accommodates 13 mm collection tubes.
  • Optimized for use with 6 cc cartridges.