Learn More
Waters Corp Test Tube Collection Rack, 13 mm, for Solid-Phase Extraction
Catalog No. 502390652 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This test tube collection rack is designed for sample collection in solid-phase extraction processes. It offers compatibility with various cartridges and test tube sizes.
  • Designed for sample collection in solid-phase extraction.
  • Compatible with 1 cc and 3 cc cartridges.
  • Accommodates 13 mm x 75 mm and 13 mm x 100 mm test tubes.
Catalog No. 50-239-0652 Supplier Waters Corp Supplier No. 186009715
This test tube collection rack is designed for sample collection in solid-phase extraction processes. It offers compatibility with various cartridges and test tube sizes.
  • Designed for sample collection in solid-phase extraction.
  • Compatible with 1 cc and 3 cc cartridges.
  • Accommodates 13 mm x 75 mm and 13 mm x 100 mm test tubes.