Learn More
Waters Corp Waters Top Rack, 3 cc, Cartridge, Otto SPEcialist Manifold
Catalog No. 502391731 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 3 cc Top Rack is an accessory designed for use with the Otto SPEcialist Positive Pressure Manifold.
  • Used with 3 cc cartridges.
  • Compatible with 13 mm collection tubes.
  • Compatible with 16 mm collection tubes.
Catalog No. 50-239-1731 Supplier Waters Corp Supplier No. 186009719
This 3 cc Top Rack is an accessory designed for use with the Otto SPEcialist Positive Pressure Manifold.
  • Used with 3 cc cartridges.
  • Compatible with 13 mm collection tubes.
  • Compatible with 16 mm collection tubes.