Learn More
Waters Corp Waters Corporation Otto Cartridge Startup Kit, 3 cc, 13 mm Tubes
Catalog No. 502391932 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Otto 3 cc Cartridge for 13 mm Collection Tubes Startup Kit is designed to facilitate the use of 3 cc flanged/flangeless cartridges with 13 mm collection tubes. This kit provides essential components for setting up a system for sample processing.
  • Includes a 3 cc top rack
  • Features a 13 mm tube collection rack
  • Contains a 13 mm tube spacer for 75 mm
  • Equipped with a drainless waste reservoir
  • Functions as an accessory stack for 24 three cc flanged or flangeless cartridges
Catalog No. 50-239-1932 Supplier Waters Corp Supplier No. 176004608
The Otto 3 cc Cartridge for 13 mm Collection Tubes Startup Kit is designed to facilitate the use of 3 cc flanged/flangeless cartridges with 13 mm collection tubes. This kit provides essential components for setting up a system for sample processing.
  • Includes a 3 cc top rack
  • Features a 13 mm tube collection rack
  • Contains a 13 mm tube spacer for 75 mm
  • Equipped with a drainless waste reservoir
  • Functions as an accessory stack for 24 three cc flanged or flangeless cartridges