Learn More
Waters Corp Waters Tube Spacer, 6 cc, 13 mm, for 75 mm
Catalog No. 502390700 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tube spacer is an accessory for the Otto SPEcialist Positive Pressure Manifold. It is designed for use with 6 cc cartridges and specifically functions as a tube spacer for 75 mm collection tubes.
  • Functions as a tube spacer for 75 mm tubes.
  • Designed for use with 6 cc cartridges.
  • Accessory for the Otto SPEcialist Positive Pressure Manifold.
  • Associated with the 6 cc cartridge for 13 mm collection tubes startup kit.
Catalog No. 50-239-0700 Supplier Waters Corp Supplier No. 186009724
This tube spacer is an accessory for the Otto SPEcialist Positive Pressure Manifold. It is designed for use with 6 cc cartridges and specifically functions as a tube spacer for 75 mm collection tubes.
  • Functions as a tube spacer for 75 mm tubes.
  • Designed for use with 6 cc cartridges.
  • Accessory for the Otto SPEcialist Positive Pressure Manifold.
  • Associated with the 6 cc cartridge for 13 mm collection tubes startup kit.